Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig Goat anti-Mouse, BB515, Polyclonal, BD

Catalog No. BDB565060 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565060 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565060 Supplier BD Biosciences Supplier No. 565060
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BD Horizon™ BB515 Goat Anti-Mouse Ig is intended to be a second-step reagent for immunofluorescent staining of cells pre-stained with Mouse Ig primary antibodies. It is reactive with the heavy and light chains of mouse primary antibodies having the IgG1, IgG2a, IgG2b, IgG3, IgM, and IgA isotypes. Minimal background staining of human and rat cells occurs in the absence of a primary mouse antibody. In addition, the BD Horizon BB515 Goat Anti-Mouse Ig stains mouse B lymphocytes with little non-specific staining of other cells. Therefore, it is useful as a primary immunofluorescent reagent for staining mouse B cells and antibody-producing cells and as a secondary reagent for staining human and rat cells pre-stained with mouse Ig primary antibodies.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Blue 515 (BB515)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Goat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.