Learn More
Innovative Research Inc Mouse IgG Affinity Purified 50mg
Supplier:  Innovative Research Inc IMSIGGAP50MG
Encompass_Preferred
Mouse IgG Affinity Purified from Innovative Research has been purified from normal Mouse Serum using Protein A affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4, with a concentration of 10.0 mg/ml and a purity of >95%.
Catalog No. 50-203-6487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.