Learn More
R&D Systems™ Mouse IgG B Cell ELISpot Development Module, 5 Plate
Shop All Novus Biologicals Products
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |