Learn More
ACROBiosystems Mouse IgG control 200tests
Supplier:  ACROBiosystems DNPAFM486200TESTS
Encompass_Preferred
APC-Labeled Mouse IgG2b Antibody Isotype Control is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a mouse monoclonal antibody with Mouse IgG2b constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.
Catalog No. 50-253-5166
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.