Learn More
ACROBiosystems Mouse IgG Fc 100ug
Supplier:  ACROBiosystems IGGS307100UG
Encompass_Preferred
Monoclonal Anti-Human-IgG-Fc Antibody Mouse IgG1 is a chimeric monoclonal antibody recombinantly expressed from CHO cells which combines the variable region of a mouse monoclonal antibody with mouse IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with IgG Fc.
Catalog No. 50-253-4624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.