Learn More
ACROBiosystems Mouse IgG Fc 100ug
Supplier:  ACROBiosystems RESA02796TESTS
Encompass_Preferred
Tumour Necrosis Factor alpha (TNF-α) ELISA Kit is based on ELISA sandwich method and designed to measure human TNF-α levels in cell culture supernates serum and plasma. It contains recombinant human TNF-α and a pair of antibodies against the recombinant factor which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human Tumour Necrosis Factor alpha (TNF-α) (ACROBiosystems cat#GMP-TNAH23) concentrations. The specificity has been verified.
Catalog No. 50-253-4245
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.