Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. HAF007 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAF007 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HAF007 Supplier R&D Systems Supplier No. HAF007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Immunohistochemistry∣Western Blot.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen IgG
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen Mouse IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects mouse IgG 1 IgG 2A IgG 2B and IgG 3 heavy and light chains in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with human IgG and rabbit IgG is observed.
Target Species Mouse
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.