Learn More
Invitrogen™ Mouse IgG Isotype Control
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against goat anti-mouse whole serum is the same as that against goat anti-mouse IgG, Fc fragment specific. No precipitin line was detected against goat anti-mouse IgA, alpha chain specific or goat anti-mouse IgM, μ chain specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Store product at 4°C until opened. After opening, dilute only enough material for a single day's use.; store remainder at 4°C under sterile conditions. Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG |
| Applications | Control, Flow Cytometry, Inhibition Assays, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Gene Alias | IgG1; Igh-4; Ighg1; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; VH7183 |
| Host Species | Mouse |
| Purification Method | Ion-exchange chromatography |
| Quantity | 5 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.