Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG Isotype Control, AF, LE, Polyclonal, Abnova™

Catalog No. 89138370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-370 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-370 Supplier Abnova Corporation Supplier No. PAB19464
Add to Cart
Add to Cart

Specifications

Antigen IgG Isotype Control
Applications ELISA, Flow Cytometry
Classification Low Endotoxin/Azide-free (LE/AF)
Concentration 0.5 mg/mL
Description Mouse IgG raised from normal mouse serum.
Dilution Flow Cytometry (≤ 1 ug/106 cells) ELISA (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Quantity 500 μg
Primary or Secondary Primary
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.