Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG Isotype Control, Texas Red, Novus Biologicals™

Mouse Antibody
Supplier: Novus Biologicals NBP197079
Description
IgG specifically detects IgG in Mouse samples. It is validated for SDS-Page.Specifications
IgG | |
Texas Red | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
SDS-Page | |
SDS-Page 1:100 - 1:2000 | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
1 mg | |
Isotype Control (Primary) | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse Serum. | |
Mouse | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction