Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Fluorescein-conjugated Antibody, R&D Systems™

Catalog No. IC002F
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC002F 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC002F Supplier R&D Systems Supplier No. IC002F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Control.

Specifications

Antigen IgG1
Classification Monoclonal
Clone 11711
Conjugate Fluorescein
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Gene Symbols IGHG1
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as mouse IgG 1 isotype control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.