Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG1 Isotype Control (CT6)

Catalog No. 01671020
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-020 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-020 Supplier Invitrogen™ Supplier No. SA135641
Add to Cart
Add to Cart

Mouse Monoclonal Isotype Control

The antibody reacts with a Guinea pig lymphocyte subset probably analog to human CD8 (cytotoxic/suppressor) subset. CD8 comprises 2 subunits, alpha and beta and exists as either an alpha/alpha homodimer or an alpha/beta heterodimer. Sequence suggests that guinea pig CD8 is more closely related to human than rat or mouse CD8. Although this monoclonal originally was developed for the detection of a Guinea pig lymphocyte subset, it also can be used as a negative control for the JSB-1 monoclonal antibodies because it is of the same IgG subclass.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1
Applications Control, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CT6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Assay Dependent
Formulation PBS with 0.1% sodium azide
Gene Alias IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C
Product Type Isotype Control
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.