Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Isotype Control (MG1) [mFluor Violet 450 SE], Novus Biologicals™
SDP

Catalog No. NB559821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB559821 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB559821 Supplier Novus Biologicals Supplier No. NBP197005MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay

Specifications

Antigen IgG1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay
Classification Monoclonal
Clone MG1
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay
Formulation 50 mM Sodium Borate
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Mouse
Purification Method Protein A purified
Quantity 0.5 mL
Regulatory Status RUO
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.