Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Isotype Control (MG1) [mFluor Violet 610 SE], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197005MFV610
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent AssaySpecifications
| IgG1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50 mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay | |
| MG1 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Protein A purified | |
| RUO | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction