Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor™ 647

Catalog No. PIMA518168
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518168 Supplier Invitrogen™ Supplier No. MA518168
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Isotype Control

This antibody This mouse IgG1 kappa monoclonal antibody (clone MOPC-21) with unknown specificity has been confirmed as a good negative control with human and rat species, based on multiple testing on rat and human tissues.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1
Applications Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MOPC-21
Concentration 1 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 15mM sodium azide
Gene Alias IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Host Species Mouse
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.