Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Isotype Control, PE, Clone: MOPC-21, Abnova™

Catalog No. 89134801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-801 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-801 Supplier Abnova Corporation Supplier No. MAB10156
Add to Cart
Add to Cart

Specifications

Antigen IgG1 Isotype Control
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MOPC-21
Conjugate PE
Description Mouse monoclonal antibody was chosen by screening on variety of resting, activated, live and fixed rat and human tissues.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Host Species Mouse
Purification Method Size-exclusion chromatography purification
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Negative reacts with human and rat.
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.