Learn More
Invitrogen™ Mouse IgG1 Isotype Control (PPV-06)
Description
This antibody reacts with undefined epitope on a plant pathogen. MA1-10405 can be used as a negative control to measure the non-specific binding of Mouse IgG1 in human, porcine, mouse and rat tissues.
Specifications
Specifications
| Antigen | Mouse IgG1 |
| Applications | Control, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | PPV-06 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene Alias | IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 |
| Host Species | Mouse |
| Immunogen | Plum pox virus. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.