Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1-kappa, FITC, Milli-Mark, Clone: MOPC-21; Isotype Control, MilliporeSigma™

Catalog No. FCMAB229FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB229FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB229FMI Supplier MilliporeSigma Supplier No. FCMAB229F
Add to Cart
Add to Cart

Mouse Isotype Control

Detect mouse IgG1 using this Milli-Mark™ Anti-mouse IgG1-FITC Antibody, clone MOPC-21 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1-kappa
Applications Flow Cytometry
Classification Monoclonal
Clone MOPC-21
Conjugate FITC
Formulation Purified mouse monoclonal IgG1κ conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Host Species Mouse
Purification Method Immunoelectrophoresis
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.