Learn More
Mouse IgG1-kappa, PE, Milli-Mark, Clone: MOPC-21; Isotype Control, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | IgG1-kappa |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MOPC-21 |
| Conjugate | PE |
| Formulation | Purified mouse monoclonal IgG1κ conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA. |
| Host Species | Mouse |
| Purification Method | Immunoelectrophoresis |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.