Learn More
Mouse IgG1 Negative Control, PE conjugate Mouse, Phycoerythrin, Clone: Ci4, Isotype Control, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | IgG1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Ci4 |
| Conjugate | PE |
| Formulation | Purified from mouse ascitic fluid. R-Phycoerythrin conjugatedready to use. The antibody is supplied in 1.0mL phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by el |
| Host Species | Mouse |
| Immunogen | Chicken immunoglobulin |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.