Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG2a Isotype Control (OX-34)

Catalog No. PIPA533238
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533238 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533238 Supplier Invitrogen™ Supplier No. PA533238
Add to Cart
Add to Cart

Mouse Monoclonal Isotype Control

PA5-33238 is suitable for use as a negative control reagent for the measurement of non-specific binding of mouse monoclonal antibodies of isotype IgG2a in human tissue. This antibody may not be suitable for intracellular staining on some cell types. The antibody recognizes a rat cell surface marker, and therefore cannot be used as a negative control in this species. This product is routinely tested in flow cytometry on rat splenocytes to confirm antibody activity and on human whole blood to test for suitability as a negative contro. Test results have shown that PA5-33238 is also suitable for use as a negative control with bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues. This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/mg. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2a
Applications Control, Flow Cytometry
Classification Monoclonal
Clone OX-34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Mouse
Immunogen Activated rat T-helper cells.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Isotype Control
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.