Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mouse IgG2a Isotype Control, Alexa Fluor™ 488
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ MG2A20

This item is not returnable.
View return policy
Description
Mouse IgG2a Isotype Control for Flow
Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
Specifications
Mouse IgG2a | |
Monoclonal | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Isotype Control | |
IgG2a | |
Wet Ice |
Flow Cytometry | |
Alexa Fluor 488 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
Liquid | |
50 Tests | |
Alexa Fluor™ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction