Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2A PerCP-conjugated Isotype Control, R&D Systems™

Catalog No. IC003C
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC003C 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC003C Supplier R&D Systems Supplier No. IC003C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Control.

Specifications

Antigen IgG2a
Classification Monoclonal
Clone 20102
Conjugate PerCP
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as mouse IgG 2A isotype control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.