Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2B Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB0043100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB0043100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB0043100 Supplier R&D Systems Supplier No. MAB0043100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG2b
Applications Flow Cytometry
Classification Monoclonal
Clone 332723
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Host Species Rat
Immunogen Cocktail of mouse IgG 2B and IgG 3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The specificity of this antibody was confirmed by direct ELISA on mouse IgG 2B monoclonal antibodies, and it does not cross-react with IgG 1 IgG 2A IgG 3 IgM, IgA, or IgE antibodies. By flow cytometry, this isotype-specific secondary antibody detects primary antibodies of the mouse IgG 2B isotype, and does not detect primary antibodies of mouse IgG 1 IgG 2A or IgG 3 class.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.