Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Negative Control Mouse, FITC, Clone: GC198, Isotype Control, MilliporeSigma™
Mouse Isotype Control
Supplier: MilliporeSigma MABC006F
Description
Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry and Immunofluorescence.
Specifications
IgG2b | |
Monoclonal | |
FITC | |
Mouse | |
1 mL | |
Store at 2 to 8°C for up to 6 months. DO NOT FREEZE. Protect from light. WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes |
|
IgG2b |
Flow Cytometry, Immunofluorescence | |
GC198 | |
Purified from mouse ascitic fluid. Fluorescein conjugated, ready to use. The antibody is supplied in 1.0mL phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by elect | |
Protein I preparation from Neisseria gonorrhoeae. | |
Primary | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction