Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2b Isotype Control, Clone: PLRV219, Abnova™

Catalog No. 89134799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-799 Supplier Abnova Corporation Supplier No. MAB10154
Add to Cart
Add to Cart

Specifications

Antigen IgG2b Isotype Control
Applications Flow Cytometry
Classification Monoclonal
Clone PLRV219
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a plant pathogen.
Dilution Flow cytometry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Purification Method Protein A purification
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Negative reacts with human.
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.