Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Isotype Control (M2B), Texas Red, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196795
Description
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG2b | |
Texas Red | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
Protein A purified | |
RUO | |
Isotyping assay resulted the material is non-reactive with antisera to mouse IgG1, IgG2a, IgG3, IgM, and IgA. Texas Red(TM) Conjugated Mouse IgG2b was proven by Dot Blot. | |
Mouse | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Flow Cytometry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
Mouse | |
0.1 mL | |
Isotype Control (Primary) | |
Reconstitute with 100 ul deionized water (or equivalent). | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction