Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Isotype Control (MPC-11), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227231UV
Description
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IgG2b | |
Monoclonal | |
DyLight 350 | |
50 mM Sodium Borate with 0.05% Sodium Azide | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
MPC-11 | |
Flow Cytometry 1 ul/1 million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Mouse | |
0.1 mL | |
Isotype Control (Primary) | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction