Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG2b kappa Isotype Control (eBMG2b), APC, eBioscience™, Invitrogen™

Catalog No. 5015168
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-151-68 25 Tests
50-112-9582 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-151-68 Supplier Invitrogen™ Supplier No. 17473241
Add to Cart
Edge
Add to Cart

Mouse Isotype Control

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: APC Mouse IgG2b Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: APC Mouse IgG2b Isotype Control is offered in 2 formats: - μg size: has been tested by flow cytometric analysis of normal human peripheral blood cells. This should be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at test size: 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2b kappa
Applications Control, Flow Cytometry
Classification Monoclonal
Clone eBMG2b
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.