Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2b Kappa Light Chain, Clone: MG2b, PerCP/Cy5.5, Isotype control, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$438.00

Specifications

Antigen IgG2b Kappa Light Chain
Clone MG2b
Dilution Flow Cytometry 1:10 - 1:1000
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14283701
SDP
View Documents
Novus Biologicals
NBP1428370.1MG
0.1 mg
Each of 1 for $438.00
Only null left
Add to Cart
 
Description

Description

IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

IgG2b Kappa Light Chain
Flow Cytometry 1:10 - 1:1000
Monoclonal
Mouse
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Primary
This monoclonal mouse IgG2b, kappa immunoglobulin is used as an isotype control for mouse IgG2b antibodies.
MG2b
Flow Cytometry
PerCP-Cyanine5.5
Mouse
IgG2b κ
Store at 4C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.