Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG3 Isotype Control (PPV-07), DyLight™ 650

Catalog No. MA1190D650
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1190D650 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1190D650 Supplier Invitrogen™ Supplier No. MA1190D650
Add to Cart
Add to Cart

Mouse Monoclonal Isotype Control

This antibody reacts with undefined epitope on a plant pathogen. MA1-190-D650 can be used as a negative control in any immunoassay where a DyLight 650-conjugated primary antibody of the mouse IgG3 isotype is used. DyLight 650 has an excitation/emission of 652/672nm.

Mouse IgG3 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the least abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG3 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG3 is less commonly used in research and diagnostic applications compared to other IgG subtypes, but it is still used in some applications such as ELISA and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG3
Applications Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PPV-07
Concentration 0.1 mg/mL
Conjugate DyLight 650
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene Alias IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3
Host Species Mouse
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.