Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgM Isotype Control (F8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221945
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Functional, In vivo assay, Block/Neutralize.Specifications
IgM | |
Monoclonal | |
LYOPH | |
Functional, Blocking/Neutralizing 50 - 200 molar excess, In vivo assay | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Protein L purified | |
RUO | |
3507 | |
Mouse | |
Purified |
Functional Assay, Blocking Assay, In vivo Assay | |
F8 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 250 ug size is provided in liquid form, PBS with No Preservative | |
Mouse | |
1 mg | |
Isotype Control (Primary) | |
Reconstitute with PBS. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction