Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgM Isotype Control, FITC, Novus Biologicals™
SDP

Mouse Antibody

Supplier:  Novus Biologicals NBP197174

Catalog No. NBP197174


Only null left
Add to Cart

Description

Description

IgM specifically detects IgM in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IgM
Fluorescein
Lyophilized from 0.06 M Potassium Phosphate, 0.45 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
IGHM
Multi-step
RUO
Isotype Control (Primary)
No reaction was observed against anti-Mouse IgG F(c). Some light chain cross reactivity will occur with anti-Mouse IgG.
Mouse
IgM
Western Blot
Western Blot
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Mouse
0.5 mg
Immunology
3507
Reconstitute with 500 ul deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only