Learn More
Description
- Also known as: NECL3; Igdf4d; Igsf4d; SynCAM2; SynCAM 2; 9330131D06; 2900078E11Rik; A830029E02Rik
- IGSF4D is a member of the nectin family which belongs to the immunoglobulin (Ig) superfamily.
- It is a transmembrane glycoprotein expressed mainly in epithelial cells and contains one Vtype Ig-like and two C2-type Ig-like domains in the extracellular region.
- These domains are characteristically responsible for Ca++ -independent homophilic and heterophilic interactions in the brain, lung, kidney, bladder, prostate and testis.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mCadm2 |
| Accession Number | NP_001334176 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1321 bp |
| Gene ID (Entrez) | 253559 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
