Learn More
Gibco™ Mouse IL-1 beta Recombinant Protein, PeproTech®
Description
211-11B-1MG will be provided as 2 x 500 μg (211-11B-500UG). Recombinant Murine IL-1beta is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P10749 |
| For Use With (Application) | Control, Western Blot Control, ELISA Standard, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 16176 |
| Molecular Weight (g/mol) | 17.5 kDa |
| Name | Mouse IL-1 beta |
| Purification Method | Purified |
| Quantity | 2 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.