Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-10, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF210101MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF210101MG 1 mg
AF21010100 100 μg
AF2101010UG 10 μg
AF21010500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF210101MG Supplier Gibco™ Supplier No. AF210101MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine IL-10 is an 18.7 kDa protein of 161 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18893
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 16153
Molecular Weight (g/mol) 18.7 kDa
Name Mouse IL-10, Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Common Name IL-10
Gene Symbol IL10
Biological Activity Determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.