Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-11 Recombinant Protein, PeproTech®

Catalog No. 220111MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
220-11-1MG 1 mg
22011100UG 100 μg
220-11-10UG 10 μg
22011250UG 250 μg
220-11-2UG 2 μg
22011500UG 500 μg
220-11-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 220-11-1MG Supplier Gibco™ Supplier No. 220111MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine IL-11 is a 19.1 kDa protein consisting of 179 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P47873
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 16156
Molecular Weight (g/mol) 19.1 kDa
Name Mouse IL-11
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11 ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin
Common Name IL-11
Gene Symbol IL11
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine T11 cells is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.