Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-12 p70 Uncoated ELISA Kit with Plates

Catalog No. 5017254
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-54 10 x 96 Tests
50-172-51 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-172-54 Supplier Invitrogen™ Supplier No. 88712186
Add to Cart
Add to Cart

Includes

Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

ELISA

The Mouse Interleukin 12 p70 (IL-12 p70) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-12 p70. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43431, P43432
Assay Range 15-2000 pg/mL
Assay Sensitivity 15 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent: 5X concentrated
Detection enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -44, -76, -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Serum, Supernatant, Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 20C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; Il-12b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; P35; p4020C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; Il-12b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; P35; p40; IL-12p35; Il-12a; Ll12a; p35;Il-12b; Il-12p40; Il12p40; p40
Gene ID (Entrez) 16159, 16160
Gene Symbol IL12A, Il12b
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2-8°C
Synonym Interleukin-12, IL12, IL23
Target IL-12 p70
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
What are the critical parameters for successful ELISPOT assay?

A: It is critically important to use a high-affinity binding PVDF membrane plate, rather than regular ELISA plate or nitrocellulose membrane plate. Additionally, highest affinity antibodies for capture work best in this assay. Empirical optimization of conditions such as cell density, stimulus/mitogen, and kinetics is necessary.

I recently bought one of your Ready-Set-Go! ELISA Sets, can I use the antibody pairs to run an ELISPOT?

We do not recommend the use of our ELISA kits for the ELISPOT application. We cannot guarantee the successful use of our ELISA kits in an ELISPOT application.

Do your Ebioscience antibody pairs work in ELISPOT as well?

Most, but not all, of our ELISA antibody pairs work for ELISPOT. We continue to evaluate and optimize our anti-human and anti-mouse cytokine antibody pairs for ELISPOT applications. Please see our protocols page (www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols) for a detailed protocol and a reference table with the list of antibody pairs that can be used in ELISPOT.

What are the components of the Ready-Set-Go! ELISPOT Sets?

Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
10X Sterile Azide-Free Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin HRP
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and enzyme

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.