Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-12 R beta 1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4951390
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1998P025 25 Tests
FAB1998P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1998P025 Supplier R&D Systems Supplier No. FAB1998P025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 1 publication

IL-12 R beta 1 Polyclonal specifically detects IL-12 R beta 1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-12 R beta 1
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q60837
Gene Alias CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Gene Symbols IL12RB1
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12 R beta 1 Val32-Glu561 Accession # Q60837
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3594
Test Specificity Detects mouse IL-12 R beta 1 in direct ELISAs and Western blots. In these formats, approximately 20% cross-reactivity with recombinant human IL-12 R beta 1 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.