Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA
SDP

Catalog No. SM1300CB Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 1 Plate)
2 Plates
6 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SM1300CB 6 Plates
M1300CB1 1 Kit (for 1 Plate)
M1300CB 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SM1300CB Supplier R&D Systems™ Supplier No. SM1300CB
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse IL-13 Culture Media Quantikine ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-13 Culture Media Quantikine ELISA quantifies mouse IL-13 in cell culture supernates (50 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 1.5pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Mouse IL-13
Sufficient For 6 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 16163
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ALRHMGC116789, BHR1interleukin-13, IL13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.