Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-13 R alpha 2 Quantikine ELISA Kit
SDP

Catalog No. M13RA2 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13RA2 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M13RA2 Supplier R&D Systems™ Supplier No. M13RA2
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure mouse IL-13 R alpha 2 levels in cell culture supernates, tissue homogenates, serum, and plasma.

The Mouse IL-13 R alpha 2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-13R alpha 2 in cell culture supernates (50 ul), tissue homogenates (13 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 39.00 - 2500pg/mL
Assay Sensitivity 5.6pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse IL-13 R alpha 2
For Use With (Application) ELISA
Gene ID (Entrez) 16165
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (13uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13 R, IL13R alpha 2, IL-13 R subunit alpha-2, IL13RA2, IL-13 RA2, IL-13 R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein
Target IL-13R alpha 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.