Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-17E (IL-25) Recombinant Protein, PeproTech®

Catalog No. 21017E500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21017E500UG 500 μg
21017E100UG 100 μg
210-17E-1MG 1 mg
21017E250UG 250 μg
21017E25UG 25 μg
210-17E-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 21017E500UG Supplier Gibco™ Supplier No. 21017E500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine IL-17E is a 35.0 kDa disulfide-linked homodimer of two 152-amino-acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8VHH8
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 140806
Molecular Weight (g/mol) 35 kDa
Name Mouse IL-17E (IL-25)
Purification Method Purified
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog
Common Name IL-25
Gene Symbol IL25
Biological Activity Measured by its ability to induce GROalpha (CXCL1) secretion in HT-29 human colon adenocarcinoma cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLRIQEGCSH LPSCCPSKEQ EPPEEWLKWS SASVSPPEPL SHTHHAESCR ASKDGPLNSR AISPWSYELD RDLNRVPQDL YHARCLCPHC VSLQTGSHMD PLGNSVPLYH NQTVFYRRPC HGEEGTHRRY CLERRLYRVS LACVCVRPRV MA
Content And Storage -20°C
Activity Measured by its ability to induce GROalpha (CXCL1) secretion in HT-29 human colon adenocarcinoma cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.