Learn More
R&D Systems™ Mouse IL-18/IL-1F4 ELISA
Catalog No. 7625
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
7625 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. 7625 Supplier R&D Systems™ Supplier No. 7625
This item is not returnable. View return policy
Mouse IL-18/IL-1F4 ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Mouse IL-18/IL-1F4 ELISA quantifies mouse IL-18/IL-1F4 in serum (30 ul).
Specifications
Product Type Mouse IL-18/IL-1F4 ELISA
Quantity 1 Kit
Storage Requirements Store unopened product at 2° to 8°C. Do not use past expiration date.