Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-1ra/IL-1F3 Quantikine ELISA Kit

Catalog No. MRA00
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MRA00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MRA00 Supplier R&D Systems™ Supplier No. MRA00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse IL-1ra/IL-1F3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-1ra/IL-1F3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-1ra/IL-1F3/IL1RN in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 13pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse IL-1 ra
For Use With (Application) ELISA
Gene ID (Entrez) 16181
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym DIRA, ICIL-1 RA, IL1F3, IL-1F3, IL-1 ra, IL1RA, IL-1 ra3, IL1RN, IL-1 RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.