Learn More
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Order Info
Ships at ambient temperature
Specifications
Specifications
| Gene | IL-20 R beta/FNDC6 |
| Accession Number | NP_001028715 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.268 |
| Insert Type | 940bp |
| Gene ID (Entrez) | 53833 |
| Gene Alias | FNDC6, DIRS1, FNDC6, IL20R beta, IL-20R2, IL-20Rb, IL20RB, IL20RB interleukin 20 receptor beta, MGC34923 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Show More |
For Reasearch Use Only. Not for use in Humans.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
