Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-22 R alpha 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066028
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4294SP 25 μg
MAB4294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4294SP Supplier R&D Systems Supplier No. MAB4294SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IL-22R alpha 1
Applications Western Blot
Classification Monoclonal
Clone 496504
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q80XZ4
Gene Alias CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Gene Symbols IL22RA1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58985
Test Specificity Detects mouse IL-22 R alpha 1 in direct ELISAs and Western blots. Shows approximately 20% cross-reactivity with recombinant human (rh) IL-22 R and no cross-reactivity with rhIL-20 R alpha , rhIL-20 R beta , or recombinant mouse IL-22BP.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.