Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-23 p19 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064509
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1619SP 25 μg
MAB1619 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1619SP Supplier R&D Systems Supplier No. MAB1619SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IL-23A/IL-23 P19 Monoclonal specifically detects IL-23A/IL-23 P19 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IL-23A/IL-23 P19
Applications Western Blot
Classification Monoclonal
Clone 320229
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P43432 (p40) & Q9EQ14 (p19)
Gene Alias IL23, IL-23, IL-23 p19, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha
Gene Symbols IL23A
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-23 p19 and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-23 heterodimers Met23-Ser335 (p40) & Leu20-Ala196 (p19) Accession # P43432 (p40) & Q9EQ14 (p19)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51561
Test Specificity Detects mouse IL-23 p19/p40 heterodimers and p19 subunits in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human p19 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.