Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IL-23 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192664 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192664 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192664 Supplier Novus Biologicals™ Supplier No. NBP192664
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Mouse IL-23R ELISA Kit (Colorimetric) measures mouse IL-23R in cell culture supernatant, serum, plasma (citrate).

Specifications

Assay Range 31.3 - 2,000pg/mL
Assay Sensitivity 16.0pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (Citrate), Serum
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 149233
Gene Symbol IL23R
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-23, Biotin-Conjugate anti-mouse IL-23 monoclonal antibody, Streptavidin-HRP, Mouse IL-23 Standard lyophilized, 4000 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Target IL-23 R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.