Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-23R Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Catalog No. p-4715319
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB16861N02 25 Tests
FAB16861N10 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB16861N02 Supplier R&D Systems Supplier No. FAB16861N025
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-23R
Applications Flow Cytometry
Classification Monoclonal
Clone 753317
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8K4B4
Gene Alias IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Gene Symbols IL23R
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149233
Test Specificity Detects endogenous mouse IL-23 R by flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.