Learn More
Description
Specifications
Specifications
| Assay Range | 62.50 to 4,000 pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sufficient For | Five 96-well plates |
| Target Species | Human |
| Gene ID (Entrez) | 146433 (Human); 76527 (Mouse); |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | Interleukin 34 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
